Orders LinkContactUsLinkComponent
Panthera Cloud Digital Microscope, Motic
Panthera Cloud Digital Microscope, Motic
Panthera cloud digital microscope
Catalog # CA76595-458
Supplier:   Motic
Supplier #:  1100403300201
Panthera Cloud Digital Microscope, Motic
Catalog # CA76595-458
Supplier:  Motic
Panthera Cloud Digital Microscope, Motic
Panthera Cloud Digital Microscope, Motic
Panthera cloud digital microscope
Price
$4,853.92
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Streamline your workflow with Panthera cloud. With the Smart CAM digital head and the ImagingOnDevice System, this SMART scope enables you to work directly from your microscope to an HDMI screen or tablet. The LAN port offers users the option to work through DigiClass or remote sharing.
  • Binocular head Siedentopf type 25° inclined, 360° rotating with SmartCam 5 MP camera
  • Built-in-digital camera: 8 MP
  • ImagingOnDevice system (Panthera App embeded software)
  • HDMI (1080i - Full HD)
  • USB 2.0 for external devices (2x)
If there's a word to define the Panthera Cloud, it is 'Smart'. It has been created to make your workflow easier, enhancing your microscopy experience. The Smart CAM digital head and its ImagingOnDevice System allows you to instantly capture wat you are viewing with the tip of your finger without the need of a computer.
Work in full HD through the HDMI port directly on a screen with the help of a mouse, or work with your tablet and the Panthera app. Store and share your discoveries easily with its unique cloud storage function. Link your panthera cloud with your cloud storage account and get ready to share your findings with anyone, wherever they are. You can also make a digital class by connecting the student's microscopes to the teacher's through the ethernet port. The possibilities are endless!
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Specifications
  • Description:
    Panthera cloud digital microscope