BA410E Compound Microscopes, Motic
About this item
The BA410E is Motic's flagship model, and it displays the highest level of optical performance. The BA410E also allows a profound diagnosis for those in pathology, haematology and cytology who require the best colour fidelity. Increased numerical apertures provide an expanded resolution power for a more detailed specimen evaluation.
- Head Siedentopf type 30° inclined, 360° rotating [trinocular: (light split = 20/80)]
- Reversed sextuple nosepiece
- Coaxial coarse and fine focusing adjustment
- Built in low position rackless stage with right hand control
- Achromat swing-out condenser N.A. 0.90 with iris diaphragm
Binocular or trinocular research microscope for diverse questions from life sciences, but also from material sciences. The standard EC-H plan achromat objectives can be replaced by fluorite and apochromatic lenses for maximum optical performance. Demanding users benefit from the system’s flexibility of the BA410E construction kit, ready to configure a personal microscope for individual requirements.
: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
2 Options Available Below
Product Details & Documents
The BA410E is Motic's flagship model, and it displays the highest level of optical performance. The BA410E also allows a profound diagnosis for those in pathology, haematology and cytology who require the best colour fidelity. Increased numerical apertures provide an expanded resolution power for a more detailed specimen evaluation.
- Head Siedentopf type 30° inclined, 360° rotating [trinocular: (light split = 20/80)]
- Reversed sextuple nosepiece
- Coaxial coarse and fine focusing adjustment
- Built in low position rackless stage with right hand control
- Achromat swing-out condenser N.A. 0.90 with iris diaphragm
Binocular or trinocular research microscope for diverse questions from life sciences, but also from material sciences. The standard EC-H plan achromat objectives can be replaced by fluorite and apochromatic lenses for maximum optical performance. Demanding users benefit from the system’s flexibility of the BA410E construction kit, ready to configure a personal microscope for individual requirements.
: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Product Family Options
Product Information
- AvailabilityPrice
- CA76595-440BA410E binocular compound microscopeSupplier Number: 1100100404301
- CA76595-442BA410E trinocular compound microscopeSupplier Number: 1100100404311