VWR® TraceClean® Low TOC Vials
About this item
These glass vial are used primarily for the collection and testing of aqueous samples for Total Organic Carbon (TOC). Secondary uses include containment of extracts. Please refer to the applicable certificate of analysis for certified analytes and corresponding reporting limits.
- Certificate of analysis and cleaning protocol form included in each product case
- Cast PTFE thin septa cap with dust covers
For pre-cleaned and certified products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement.
D10 protocol is a depyrognation-like process for low level Total Organic Carbon (TOC). Caps are 1-piece Open top polypropylene with cast PTFE septa ultrasonically bonded (not glued); overall thickness 0.060 ±0.005" (silicone= 0.056", PTFE= 0.004"). The PTFE liner is a 0.10" thick disc.
Certifications: Meets ASTM standards. EN 342.
2 Options Available Below
Product Details & Documents
These glass vial are used primarily for the collection and testing of aqueous samples for Total Organic Carbon (TOC). Secondary uses include containment of extracts. Please refer to the applicable certificate of analysis for certified analytes and corresponding reporting limits.
- Certificate of analysis and cleaning protocol form included in each product case
- Cast PTFE thin septa cap with dust covers
For pre-cleaned and certified products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement.
D10 protocol is a depyrognation-like process for low level Total Organic Carbon (TOC). Caps are 1-piece Open top polypropylene with cast PTFE septa ultrasonically bonded (not glued); overall thickness 0.060 ±0.005" (silicone= 0.056", PTFE= 0.004"). The PTFE liner is a 0.10" thick disc.
Certifications: Meets ASTM standards. EN 342.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- CapacityDiameter ×HColorCap includedCap typeCap SizeCap colorSeptumAvailabilityPrice
- 76423-174TOC vials, <10 ppb TOCTOC vials, <10 ppb TOC40.00 ml27.5×95 mmClearYesOpen top, 1-piece24-414WhiteCast thin PTFELog in for availabilityCase of 80$352.24
- 76423-176TOC vials, <20 ppb TOCTOC vials, <20 ppb TOC40.00 ml27.5×95 mmClearYesOpen top, 1-piece24-414WhiteCast thin PTFELog in for availabilityCase of 80$265.99
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.