To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Calibration:Up to 3 points
- Temperature range:0...50 °C (32...122 °F)
- Description:Oakton® PCTSTestr™ 30 pH/conductivity/TDS/salinity waterproof pocket tester
- Environmentally Preferable:
- Temperature accuracy:±1.0 °C (±1.8 °F)
- Temperature resolution:0.1 °C/°F
- Battery life:200 hours
- Parameters:pH/Conductivity/TDS/Salinity
- Automatic buffer recognition:NIST and USA
- Conductivity accuracy:±1% full scale
- Conductivity range:0.00 - 20.00 mS/cm
- Conductivity resolution:0.1 µS/cm, 1 µS/cm, 0.01 mS/cm
- Display:Dual-line LCD
- Salinity accuracy:±1% full scale
- Salinity range:0.1 - 10.0 ppt
- Salinity resolution:0.1 ppt
- TDS accuracy:±1% full scale
- TDS range:0.1 - 10.0 ppt
- TDS resolution:0.1 ppm, 1 ppm, 0.01 ppt
- Temperature compensation:Automatic
- W×D×H:38.1×177.8×38.1 mm (1.5×7×1.5")
- pH accuracy:±0.01 pH, ±1 digit
- pH range:−2 - +16
- pH resolution:0.01
- Cat. no.:76595-384
Specifications
About this item
Ultimate tester for pH, conductivity, TDS, salinity and temperature.
- Combine measurement capabilities of three testers in one convenient pocket-sized unit
- Up to 3-point pH calibration with NIST or USA auto buffer recognition
- Conductivity, TDS, and salinity feature auto ranging based on the concentration of your sample
- Redesigned leakproof cap functions as a sample, calibration, and hydrating cup
- Auto-shutoff function conserves battery life
- IP67-rated housing is waterproof and floats when sensor cap is on
Great for water, wastewater, laboratory or plant use, the Oakton® PCTS 30 multiparameter tester features long-life pH electrodes with a wide range of sample compatibility. Pin-style conductivity sensors feature stainless steel electrodes for chemical resistance and durability. Replaceable sensor module saves you money by reusing the meter body.
The double junction tester helps you bring the performance of your lab instruments out into the field. Features include large easy-to-read LCD with 2-color backlight, user-adjustable temperature coefficient, self-adjusting TDS and salinity factors, and low-battery indicator. Leakproof cap provides stabilizing base to stand the meter upright.