Heated Forceps, Mopec
About this item
Mopec’s heated forceps are electrically heated instruments designed to assist with the proper positioning of tissue specimens during the embedding process in paraffin wax. The safe, low-voltage heating element maintains a steady temperature throughout specimen manipulation.
- Designed to take up minimal bench-top space
- Anti-microbial coating to help prevent bacteria and virus growth
- Molded finger grips increase user comfort and remain cool when in use
- Bayonet connection prevents disconnection while in use
The ergonomic design ensures user comfort while performing specimen placement, and the anti-microbial coating helps prevent bacteria and virus growth. Choose between serrated or smooth tips. Available in three sizes and compatible with the Mopec control unit.
4 Options Available Below
Product Details & Documents
Mopec’s heated forceps are electrically heated instruments designed to assist with the proper positioning of tissue specimens during the embedding process in paraffin wax. The safe, low-voltage heating element maintains a steady temperature throughout specimen manipulation.
- Designed to take up minimal bench-top space
- Anti-microbial coating to help prevent bacteria and virus growth
- Molded finger grips increase user comfort and remain cool when in use
- Bayonet connection prevents disconnection while in use
The ergonomic design ensures user comfort while performing specimen placement, and the anti-microbial coating helps prevent bacteria and virus growth. Choose between serrated or smooth tips. Available in three sizes and compatible with the Mopec control unit.
Product Family Options
Product Information
- ColorTip styleTip widthAvailabilityPrice
- 76579-226Heated forcepsSupplier Number: BK301-SE
- 76579-232Heated forcepsSupplier Number: BK301-SM
- 76579-228Heated forcepsSupplier Number: BK302-SE
- 76579-234Heated forcepsHeated forcepsYellowSmooth2 mmDiscontinued-Supplier Number: BK302-SMSpecifications:MarketsourceMarketsource
- 76579-230Heated forcepsSupplier Number: BK304-SE
- 76579-236Heated forcepsHeated forcepsBlueSmooth4 mmDiscontinued-Supplier Number: BK304-SMSpecifications:MarketsourceMarketsource
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)