Orders LinkContactUsLinkComponent
Slide Drying Mini Workstation
Slide Drying Mini Workstation
Slide drying mini workstation, 110 or 230 V
Catalog # 76579-218
Supplier:   Mopec
Supplier #:  BK225
Slide Drying Mini Workstation
Catalog # 76579-218
Supplier:  Mopec
Slide Drying Mini Workstation
Slide Drying Mini Workstation
Slide drying mini workstation, 110 or 230 V
Price
4 005,18 $
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mopec’s slide drying ,mini workstation for flotation baths was specifically designed to fit directly over our illuminated tissue flotation bath and take up minimal bench-top space. This workstation features a mini hotplate to remove wrinkles from tissue sections and a sloped drying area that holds up to 12 slides.
  • Dual Voltage: 110 and 230 V power supplies
    Designed to take up minimal bench-top space
  • Fits directly over our illuminated tissue flotation bath
  • Digital temperature control
  • Rapid heating system ensures system is ready to use in less time
  • Mini hotplate removes wrinkles from tissue sections
  • Sloped drying area improves drying time and can hold up to 12 slides
  • Anti-microbial coating to help prevent bacteria and virus growth
Sold individually.
Specifications
  • Description:
    Slide drying mini workstation
  • Electrical:
    110 or 230 V
  • Cat. No.:
    76579-218
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)