If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Biohazard waste bags
- Cat. no.:76579-394
Specifications
About this item
Mopec’s biohazard waste bags are made of a durable, autoclavable, 2.25 HDPE material and are clearly marked with the 'Biohazard' symbol.
- Made from a durable, autoclavable, 2.25 HDPE material
- Dimensions: 25×35"
- Capacity: 12 to 16 galloons
- Can withstand temperatures up to 250 ˚F during autoclaving
- Feature OSHA-standard 'Biohazard' symbol and words 'Hazardous Waste'
- Color-changing sterilization indicator
- Features large, white marking panel for use with permanent markers
- Fits perfectly into Biohazard Waste Bin (BG681-B, sold separately)
Bags feature drawstring loops, these bags allow for proper ventilation during sterilization and provide the user with a safe and easy way to remove bags from a hot autoclave. Capacity of 12 to 16 gallons. Sold by the case of 100 bags.