Orders LinkContactUsLinkComponent
Illuminated Tissue Flotation Bath
Illuminated Tissue Flotation Bath
Tissue flotation bath, 110 or 230 V
Catalog # 76579-214
Supplier:   Mopec
Supplier #:  BK200
Illuminated Tissue Flotation Bath
Catalog # 76579-214
Supplier:  Mopec
Illuminated Tissue Flotation Bath
Illuminated Tissue Flotation Bath
Tissue flotation bath, 110 or 230 V
Price
$4,272.03
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mopec’s Illuminated Tissue Flotation Bath features an LED light and high-contrast background that allows users to better view tissue sections while also detecting foreign particles floating on the surface of the water. The LED light is dimmer controlled and can be switched between white or blue.
  • Dual Voltage: 110 and 230 V power supplies
    Designed to take up minimal bench-top space
  • Switch between blue or white LED back lighting with dimmer control for optimal viewing
  • Extra deep bowl to assist with tissue pick-up at any angle
  • Digital temperature control
  • Rapid heating system ensures system is ready to use in less time
  • Convenient slide drying area at the rear of the bath
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Optional lid helps improve temperature control and prevent surface particles (76579-216)
Specifications
  • Description:
    Tissue flotation bath, 110 or 230 V
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)