Orders LinkContactUsLinkComponent
Round Tissue Flotation Bath
Round Tissue Flotation Bath
Round tissue flotation bath, 110 or 230 V
Catalog # 76579-210
Supplier:   Mopec
Supplier #:  BK100
Round Tissue Flotation Bath
Catalog # 76579-210
Supplier:  Mopec
Round Tissue Flotation Bath
Round Tissue Flotation Bath
Round tissue flotation bath, 110 or 230 V
Price
$2,404.07
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mopec’s Round Tissue Flotation Bath is designed with a deep center bowl so slides can be fully submerged at a convenient angle. It features an anti-microbial coating to protect the system from bacterial and viral growth as well as a rapid heating system to reduce waiting time.
  • Dual Voltage: 110 and 230 V power supplies
    Designed to take up minimal bench-top space
  • Deep bowl to assist with tissue pick-up at any angle
  • Rapid heating system ensures system is ready to use in less time
  • Analog temperature control
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Optional lid helps improve temperature control and prevent surface particles (76579-212)
Specifications
  • Description:
    Round tissue flotation bath, 110 or 230 V
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)