ContactUsLinkComponent
Klercide™ mop wipe frame, stainless steel
Klercide™ mop wipe frame, stainless steel
Catalog # 76554-276
undefined
Klercide™ mop wipe frame, stainless steel
Catalog # 76554-276
Supplier Number:  10056359
Specifications
  • Description:
    Mop wipe frame, incl. one head and one handle
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Cat. no.:
    76554-276
Specifications
About this item
This stainless steel mop frame features a stiff movable flat top.
  • Fully autoclavable
  • Made from stainless steel with stiff, moveable flat mop head frame
  • Designed for use with Klercide™ pre-saturated mop wipes
  • Supplied Gamma irradiated
Option of adding Klercide™ silicone pad for better contact on uneven surfaces.