Orders LinkContactUsLinkComponent
Rapid Response™ Lateral Flow Reader, BTNX
Rapid Response™ lateral flow reader
Catalog # 76547-874
Supplier:   BTNX INC.
 :  LFA-1.0
Rapid Response™ Lateral Flow Reader, BTNX
Catalog # 76547-874
Supplier:  BTNX INC.
Rapid Response™ Lateral Flow Reader, BTNX
Rapid Response™ Lateral Flow Reader, BTNX
Rapid Response™ lateral flow reader
Price
$2,193.19
Log in for availability
Each Case of 8
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The Rapid Response™ lateral flow reader analyzes results and automatically generates and stores test reports from Rapid Response™ Multi-drug test panels.
  • Integrates directly with PateintVuTM and other LIS and EMR systems for seamless workflow, administration and record-keeping
  • Analyzes results within 10 s
  • Saves test results and generates customized test results
  • QR code recognition identifies the patient, sample and test type
  • Flexible operator access, test and method settings
This product is only compatible with Rapid Response™ Multi-drug test panels.
Specifications
  • Description:
    Rapid Response™ lateral flow reader