Dog ApolipoProtein B (APOB) ELISA Kit, AFG Bioscience
76578-846
:
- Assay Type:Sandwich
- Host:Dog
- Target protein:Apolipoprotein B
- Description:Dog Apolipoprotein B (APOB) ELISA kit
- Size:96 Wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:200 - 6400 ng/ml
- Storage temperature:2…8 °C
- Sample volume:50 - 100 µl
- Sensitivity:23.44 ng/ml
- Regulatory status:RUO
- Cat. no.:76578-846
- Assay Principle: Quantitative
- Research Topic: Cancer ,Cardiovascular
- Abbreviation: APOB
- Detection Wavelength: 450 nm