Bovine Pentraxin-related Protein PTX3 (PTX3) ELISA Kit, AFG Bioscience
76578-402
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Assay Type:Sandwich
- Host:Bovine
- Target protein:Pentraxin-related protein PTX3
- Description:Bovine Pentraxin-related protein PTX3(PTX3) ELISA kit
- Size:96 Wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:5 - 150 ng/ml
- Storage temperature:2…8 °C
- Sample volume:50 - 100 µl
- Sensitivity:1 ng/ml
- Regulatory status:RUO
- Cat. no.:76578-402
- Assay Principle: Quantitative
- Research Topic: Cardiovascular
- Abbreviation: PTX3
- Detection Wavelength: 450 nm