Order Entry
Canada
ContactUsLinkComponent
Bovine Matrix Metalloproteinase-20 (MMP20) ELISA Kit, AFG Bioscience
Bovine Matrix Metalloproteinase-20 (MMP20) ELISA Kit, AFG Bioscience
Catalog # 76576-180
Supplier:  AFG Bioscience
CAS Number:  
undefined
Bovine Matrix Metalloproteinase-20 (MMP20) ELISA Kit, AFG Bioscience
Catalog # 76576-180
Supplier:  AFG Bioscience
Supplier Number:  EK760156
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Sandwich
  • Host:
    Bovine
  • Target protein:
    Matrix metalloproteinase-20
  • Description:
    Bovine Matrix metalloproteinase-20(MMP20) ELISA kit
  • Size:
    96 Wells
  • Sample type:
    Serum, plasma, urine, tissue homogenates, cell culture supernates
  • Time to Results:
    1 - 5 h
  • Shelf life:
    6 months (4 °C); 12 months (–20 °C)
  • Detection range:
  • Storage temperature:
    2…8 °C
  • Sample volume:
    50 - 100 µl
  • Sensitivity:
  • Regulatory status:
    RUO
  • Cat. no.:
    76576-180

Specifications

About this item

  • Research Topic: Cancer
  • Abbreviation: MMP20
  • Detection Wavelength: 450 nm