Bovine Endothelin-Converting Enzyme 1 (ECE1) ELISA Kit, AFG Bioscience
Catalog # 76576-990
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Bovine
- Target protein:Endothelin-converting enzyme 1
- Description:Bovine Endothelin-converting enzyme 1(ECE1) ELISA kit
- Size:96 Wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:—
- Storage temperature:2…8 °C
- Sample volume:50 - 100 µl
- Sensitivity:—
- Regulatory status:RUO
- Cat. no.:76576-990
Specifications
About this item
- Research Topic: Cancer
- Abbreviation: ECE1
- Detection Wavelength: 450 nm