Human Interleukin-24 (IL-24) ELISA Kit, AFG Bioscience
Catalogue # 76576-988
Fournisseur: AFG Bioscience
Numéro CAS:
Spécifications
- Assay Type:Sandwich
- Host:Human
- Target protein:Interleukin-24
- Description:Human Interleukin-24 (IL-24) ELISA kit
- Size:96 Wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:1 - 100 pg/ml
- Storage temperature:2…8 °C
- Sample volume:50 - 100 µl
- Sensitivity:0.2 pg/ml
- Regulatory status:RUO
- Cat. no.:76576-988
Spécifications
À propos de cet article
- Assay principle: Quantitative
- Research topic: Immunology
- Abbreviation: IL-24
- Detection wavelength: 450 nm