Crucibles, Wide Form with Cover, Porcelain, United Scientific Supplies
Supplier: United Scientific Supplies
Certificates
About this item
Wide form crucibles include covers. Crucibles are glazed on inner and outer surfaces except for the outside bottom and rim.
- Autoclavable
- Withstand temperatures to 1150 °C
To prevent thermal stress cracks, we strongly recommend a heating/cooling rate not exceeding 200 °C per hour. United porcelain crucibles offer excellent resistance to acids and alkalies except for hydrofluoric acid.
Frequently Bought Together