Eisco Test Sieves/Strainers, Set of 6, ASTM
Catalog # 470344-206
Supplier: Wards
Specifications
- Description:Test sieves
- Cat. no.:470344-206
Specifications
About this item
Plastic frames with stainless steel mesh for serial sifting of samples.
- Six plastic sieves with stainless steel mesh, cover and bottom collector
- Complete with plastic lid and bottom collector
- Stackable
Each sieve has a plastic frame with a single mesh size (ASTM mesh no. 5, 10, 35, 60, 120, 230). Total height of stacked sieves: 15.25". Not foodsafe.