Eisco Mountable Wooden Draining Rack
470344-132
: Wards
- Description:Draining rack
- Cat. no.:470344-132
- No. of Places:90
Holds up to 90 labware pieces.
- Sturdy polished beech wood construction will stand up to the wear and tear of the lab
- Equipped with 2 metal hang tabs for easy wall mounting
Nine tiers of pegs include 18 draining pins measuring 5" in length, and 72 pins measuring 3" in length. Entire rack measures 20" wide, 5.5" long and 20" tall. Rack can be mounted to wall via two metal hang tabs (screws not included).