ContactUsLinkComponent
Innovating Science® Chromatography of Amino Acids
Innovating Science® Chromatography of Amino Acids
Catalog # 470343-450
Supplier:  Ward's Science
undefined
Innovating Science® Chromatography of Amino Acids
Catalog # 470343-450
Supplier:  Ward's Science
Supplier Number:  IS2522
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Chromatography of amino acids, full kit
  • Cat. no.:
    470343-450
  • No. of Uses:
    15
Specifications
About this item
Learn about amino acids, the building blocks of proteins, and how to separate and identify them using paper chromatography.
  • Students will perform a paper chromatography experiment on 3 known amino acids, and then use their results to identify the components of an unknown amino acid mixture.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2, HS-PS1-3
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include planning and carrying out investigations, analyzing and interpreting data, scientific knowledge is based on empirical evidence
Amino acids are the fundamental building blocks of all proteins. Often times scientists may need to identify the amino acid contained in a protein. One such method for separating and identifying individual amino acids is paper chromatography. In this activity, students will perform a paper chromatography experiment on three known amino acids, and then use their results to identify the components of an unknown amino acid mixture. This kit contains enough material for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery: Instructor's manual and SDS included.
Caution: This product can expose you to chemicals including Methanol/Methyl isobutyl ketone, which is known to the State of California to cause cancer and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.