ContactUsLinkComponent
Innovating Science® Physical and Chemical Properties of Matter
Innovating Science® Physical and Chemical Properties of Matter
Catalog # 470343-444
Supplier:  Ward's Science
undefined
Innovating Science® Physical and Chemical Properties of Matter
Catalog # 470343-444
Supplier:  Ward's Science
Supplier Number:  IS2517
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Physical and chemical properties of matter
  • Cat. no.:
    470343-444
  • No. of Uses:
    10
Specifications
About this item
Learn about matter and the physical and chemical properties of matter.
  • Students will perform a series of experiments on density, smell, color, melting point, boiling point, and solubility, and then perform a series of reactions to try to identify an unknown solution with what they learned.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2, HS-PS1-1, HS-PS1-3
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data, developing and using models, planning and carrying out investigations
The activities in this kit focus on developing a better understanding of what is matter and what the different types of properties are. Students perform a series of experiments focusing on the physical properties of density, smell, color, melting point, boiling point and solubility. Other experiments focus on chemical properties in which students perform a series of reactions as they try and identify an unknown solution. Kit contains enough material for 10 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery: Kit includes instructional manual and SDS booklet.
Caution: This product can expose you to chemicals including Respirable crystalline silica, which are known to the State of California to cause cancer. For more information go to http://www.P65Warnings.ca.gov.