ContactUsLinkComponent
Innovating Science® Organic Chemistry: Chemical Identification of Biomolecules
Innovating Science® Organic Chemistry: Chemical Identification of Biomolecules
Catalog # 470343-440
Supplier:  Ward's Science
undefined
Innovating Science® Organic Chemistry: Chemical Identification of Biomolecules
Catalog # 470343-440
Supplier:  Ward's Science
Supplier Number:  IS2508
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Organic chemistry: chemical identification of biomolecules
  • Cat. no.:
    470343-440
  • No. of Uses:
    15
Specifications
About this item
Learn about the importance of proteins, carbohydrates, and lipids in living organisms.
  • Students will learn to test for presence of proteins, sugars, starch, and lipids.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data
Understand the importance of proteins, carbohydrates, and lipids in living organisms. Students will learn to identify a positive test result for proteins using biuret reagent, examine the reaction between Benedict's reagent and a simple sugar, use iodine/potassium iodide to test for the presence of starch, and test for the presence of lipids using a fat-soluble dye. Kit contains enough materials for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery: Kit includes instructional manual and SDS booklet.