ContactUsLinkComponent
Innovating Science® Density and Displacement
Innovating Science® Density and Displacement
Catalog # 470343-424
Supplier:  Ward's Science
undefined
Innovating Science® Density and Displacement
Catalog # 470343-424
Supplier:  Ward's Science
Supplier Number:  IS2001
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Density and displacement
  • Cat. no.:
    470343-424
  • No. of Uses:
    15
Specifications
About this item
Learn how to determine density and displacement.
  • Students will determine the density of four metals by determining their mass and displacement
  • Aligned to NGSS Practices
In this exercise, you will determine the density of four metals, similar in appearance. Mass will be determined using an Ohaus Scout® Pro electronic balance and volume will be calculated using Archimedes Principle of Displacement. The data obtained will then be compared to published density values. Kit contains enough materials for 15 groups.
Delivery: Kit includes instructional manual and SDS booklet.
Caution: This product can expose you to chemicals including Lead and lead compounds and Nickel metallic, which are known to the State of California to cause cancer and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.