ContactUsLinkComponent
Innovating Science® Presumptive Gun Shot Residue Kit
Innovating Science® Presumptive Gun Shot Residue Kit
Catalog # 470343-392
Supplier:  Ward's Science
undefined
Innovating Science® Presumptive Gun Shot Residue Kit
Catalog # 470343-392
Supplier:  Ward's Science
Supplier Number:  IS9001
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Presumptive gun shot residue kit
  • Cat. no.:
    470343-392
  • No. of Uses:
    30
Specifications
About this item
Learn about how to test if a surface has been exposed to gunpowder residue.
  • Students will perform a two part test to determine whether a surface has been exposed to a discharged firearm
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data
This is a two part test to determine whether a surface has been exposed to a discharged firearm. A rapid color change takes place to verify the presence of nitrates and lead. Kit contains instructions and enough materials for 30 tests.
Delivery: Kit includes instructional manual and SDS booklet.
Caution: This product can expose you to chemicals including Strong inorganic acid mists containing sulfuric acid, Lead and lead compounds, which are known to the State of California to cause cancer and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.