ContactUsLinkComponent
Innovating Science® Anesthefly Kit
Innovating Science® Anesthefly Kit
Catalog # 470343-474
Supplier:  Ward's Science
undefined
Innovating Science® Anesthefly Kit
Catalog # 470343-474
Supplier:  Ward's Science
Supplier Number:  IS3005
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Anesthefly kit
  • Cat. no.:
    470343-474
  • No. of Uses:
    5
Specifications
About this item
Learn how to anesthetize small insects without killing or sterilizing.
  • Students will anesthetize Drosophila melanogaster for 40 to 50 minutes
  • Aligned to NGSS Practices
Anesthetize Drosophila melanogaster fruit fly and other small insects for at least 40 to 50 minutes without killing or sterilizing. Includes instructions.
Delivery: Kit includes instructional manual and SDS booklet.
Caution: This product can expose you to chemicals including Methanol/Methyl isobutyl ketone, which is known to the State of California to cause cancer and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.