Acetone ≥99.9% (dry basis) for HPLC, not for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
BJCS010-200
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Pack type:Stainless steel drum, returnable
- Size:200 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent selection guide recommended after discussion
- Cat. no.:BJCS010-200
- Supplier No.:CS010-200
- GC Analysis (excluding water):99.9 % min.
- Refractive Index (20ºC):1.3583 - 1.3589
- Residue:1 mg/L max.
- UV Absorbance @ 340 nm:0.06 AU max.
- UV Absorbance @ 350 nm:0.01 AU max.
- UV Absorbance @ 375 nm:0.005 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:330 nm max.
- Water by Karl Fischer Titration:0.5 % max.
:
:58.08 g/mol
:56 °C
:–94.8 °C
:0.79 g/cm³ (20 °C)
:–20 °C
:1090
:3,II
:GHS07
:GHS02
:Ambient
:67-64-1
:CH₃COCH₃
:MFCD00008765