Bond Elut PBA Plates, Agilent Technologies
Catalog # CAAGA496121C
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Volume:1 ml
- Description:VersaPlate well plate tubes, NH2, 50 mg, 40 µm, PE frit (20 µm)
- Environmentally Preferable:
- Cat. no.:CAAGA496121C
Specifications
About this item
Bond Elut PBA uses phenylboronic acid functionality to retain analytes.
- Normal phase or anion-exchange sorbent
- Weaker anion exchange than SAX
- Amenable to separating structural isomers
Bond Elut PBA is a unique silica SPE sorbent containing a phenylboronic acid functionality that can retain analytes through a reversible covalent bond. This strong covalent retention mechanism enables high specificity and cleanliness. The boronate group has a strong affinity for cis-diol-containing compounds, such as catechols and nucleic acids.