Order Entry
Canada
ContactUsLinkComponent
Well Plate Tubes for Bond Elut LMS Plates, Agilent Technologies
Well Plate Tubes for Bond Elut LMS Plates, Agilent Technologies
Catalogue # CAAG75510050
Well Plate Tubes for Bond Elut LMS Plates, Agilent Technologies
Catalogue # CAAG75510050
Supplier Number:  75510050

Spécifications

  • Volume:
    1.8 ml
  • Description:
    VersaPlate well plate tubes, LMS, 50 mg, PE frit (20 µm)
  • Cat. no.:
    CAAG75510050

Spécifications

À propos de cet article

Bond Elut LMS enables elution without the secondary reactions of additives.

  • Ultraclean styrene-divinylbenzene polymer
  • Optimized 75 µm particle size for reproducible flow
  • High capacity and surface area for efficient extraction
  • Large pore size (300 Å) for large molecule extraction

Bond Elut LMS (large molecule size) polymeric sorbent lets you elute without having to add amine modifiers, buffers, or acids. The elimination of secondary interactions means that analytes can be eluted with pure organic solvents or HPLC-compatible solvent mixtures of low ionic strength, crucial for compatibility with LC/MS or other delicate analytical techniques.