Bond Elut LMS Plates, Agilent Technologies
Catalog # CAAGA3961025
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Volume:2 ml
- Description:Bond Elut LMS, 96 square-well PP plate, 25 mg, PE frit (20 µm), full skirt
- Cat. no.:CAAGA3961025
Specifications
About this item
Bond Elut LMS enables elution without the secondary reactions of additives.
- Ultraclean styrene-divinylbenzene polymer
- Optimized 75 µm particle size for reproducible flow
- High capacity and surface area for efficient extraction
- Large pore size (300 Å) for large molecule extraction
Bond Elut LMS (large molecule size) polymeric sorbent lets you elute without having to add amine modifiers, buffers, or acids. The elimination of secondary interactions means that analytes can be eluted with pure organic solvents or HPLC-compatible solvent mixtures of low ionic strength, crucial for compatibility with LC/MS or other delicate analytical techniques.