Bond Elut C18 EWP, Agilent Technologies
Catalog #: CAAG12256130
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Capacity:6 ml
- Column Format:Cartridge
- Film thickness:40.00 µm
- Particle size:40 µm
- Description:Bond Elut straight barrel cartridge, PE frit (20 µm)
- Temperature limit:Apolar stationary phase
- Bed Weight:1 g
- Column Hardware:PP
- Packing Material:C18 EWP
- Phase:PP
Specifications
About this item
Bond Elut C18 EWP features wide pores, making it ideal for efficient extraction of macromolecules.
- No exclusion of large molecules
- Good for desalting proteins
- Successful separation of proteins
- peptides, or nucleotides
Bond Elut EWP is based upon standard particle size silica but with 500 Å pores to allow more efficient extraction of large molecules (>15,000 MW), which are typically excluded from standard porosity silica phases. Typical matrices include aqueous samples and biological fluids.
Customers Who Bought This Also Bought