$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Tubes
Test Tubes
Well Plate Tubes for Bond Elut Plexa PCX Plates, Agilent Technologies
Well Plate Tubes for Bond Elut Plexa PCX Plates, Agilent Technologies
CAAG75580060
:
AGILENT TECHNOLOGIES, INC (CSD) CA
Well Plate Tubes for Bond Elut Plexa PCX Plates, Agilent Technologies
CAAG75580060
:
AGILENT TECHNOLOGIES, INC (CSD) CA
:
75580060
Volume:
1.8 ml
Description:
VersaPlate well plate tubes, Plexa PCX, 60 mg, PE frit (20 µm)
Cat. no.:
CAAG75580060