Well Plate Vacuum Manifolds, Agilent Technologies
Catalog #: CAAG5133000
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Well Plate Vacuum Manifolds, Agilent Technologies
Catalog #: CAAG5133000
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:96-well plate vacuum manifold, acrylic
Specifications
About this item
Customers Who Bought This Also Bought
Sodium hydroxide 40% (w/v) in aqueous solution for Kjeldahl nitrogen analysis nitrogen-free
Supplier: Ricca Chemical
670,90 $
Masterflex® L/S® Easy-Load® II Pump Heads for Precision Tubing, Avantor®
Supplier: Avantor Fluid Handling
1 039,64 $
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
624,91 $
Buffer Solution, pH 7.00 (±0.01 @ 25 °C), No Color, Specpure®, NIST Traceable, Thermo Scientific
Supplier: Thermo Scientific Chemicals
56,00 $