Argon Humidifier and Supplies for ICP-OES, Agilent Technologies
Catalog # CAAGG840060710
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Tubing set, for argon humidifier, for Agilent ICP-MS,ICP-OES and MP-AES
- Cat. no.:CAAGG840060710
Specifications
About this item
Conquer your most challenging samples with the ICP-OES argon humudifier accessory.
- Achieve accurate, reproducible analysis of samples containing elevated levels of total dissolved solids
- Enables the accurate analysis of challenging sample types that may potentially cause particulate buildup, and eventually block the nebulizer
- Reduces frequency of routine maintenance when analyzing samples with elevated TDS
- Significantly improve long-term stability for analysis of challenging samples
The Agilent argon humidifier accessory is compatible with Agilent 5100/5110/5800 and 5900 ICP-OES. The accessory is based on hollow fiber membrane tubing, which passes through a bottle containing de-ionized water. This humidifies the nebulizer gas, reducing desolvation at the nebulizer orifice. This process enables the accurate analysis of challenging sample types that could cause particulate buildup, and eventually block the nebulizer, improving long term stability. Reccomended when solution have >3% TDS.