Spray Chambers and Supplies for PerkinElmer ICP-MS Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Glass cyclonic, inert PFA spray chambers and supplies for PerkinElmer ELAN and NexION ICP-MS instuments.
- Guaranteed to be compatible with PerkinElmer ICP-MS instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Qualified on a genuine PerkinElmer system
- Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
- Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns, to help you get the most out of your PerkinElmer system
- In stock and available worldwide for 24- to 48-hour delivery (in most regions)
- Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
- Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent glass cyclonic, inert PFA spray chambers and supplies are equivalent to the PerkinElmer ICP-MS versions. These parts provide the best precision, efficient drainage, and reduced interferences and you can use them with complete confidence.
Use the single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout. Alternatively, the double pass glass cyclonic spray chamber provides improved plasma stability with organic solvents and samples that have high levels of total dissolved solids.
Frequently Bought Together