Miscellaneous Supplies for MP-AES, Agilent Technologies
Catalog # CAAGG800064112
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Pre-optic window for Agilent MP-AES
- Cat. no.:CAAGG800064112
Specifications
About this item
Supplies for MP-AES include Pre-optic window and disposable Onboard Argon bottle for plasma ignition.
- Miscellaneous supplies can help you to maintain the superior performance of the MP-AES
- Easily order for your planned maintenance and ensuring your supplies cabinet is properly provisioned
Agilent can help you to maintain the superior performance of the MP-AES with all the supplies and accessories you need. Miscellaneous supplies include user replaceable Pre-optic window for MP-AES, and disposable Onboard Argon bottle for plasma ignition (not available in all countries).