ContactUsLinkComponent
Mopec Safe Seal System, Single Facility Pack, Mortech
Mopec Safe Seal System, Single Facility Pack, Mortech
Catalog # 76549-654
Supplier:  Mopec
undefined
Mopec Safe Seal System, Single Facility Pack, Mortech
Catalog # 76549-654
Supplier:  Mopec
Supplier Number:  BE163
Specifications
  • Dimensions:
    Mopec Safe Seal System, single facility pack
  • Cat. no.:
    76549-654
Specifications
About this item
Mopec Safe Seal System is a military tested, heavy duty, hermetically sealable Bio-Safety Level 4 (BSL-4) Containment body bag system. It is easy to use, as the body is placed in the flexible material, then heat-sealed with the Safe Seal Thermosealer to provide complete and secure containment of infectious human or animal remains. The Safe Seal System was designed to help keep workers and survivors safe after a mass fatality event or any high-risk biohazard exposure.
Delivery: The Facility Pack contains 1 roll of Mopec Safe Seal (BE162), 1 Heat Sealer (BE153), 1 Vertical Stand (BE152), 2 Alignment Clips (BE900), 1 Razor Knife (BE901), and 1 Extension Cord (BE902).