Order Entry
Canada
Orders LinkContactUsLinkComponent
Funnel With Attached Screen, Mortech

Funnel With Attached Screen, Mortech

136.5 mm (5⅜"), 127 mm (5")
Catalog # 76498-238
Supplier:   Mopec
Supplier #:  BA077
Funnel With Attached Screen, Mortech
Catalog # 76498-238
Supplier:  Mopec
Funnel With Attached Screen, Mortech
Funnel With Attached Screen, Mortech

136.5 mm (5⅜"), 127 mm (5")

Price
$328.87
Log in for availability
Each
Product Details & Documents

Stainless steel funnel and screen used for formalin collection systems and specimen pour-off.

  • 20 gauge stainless steel and wire mesh
Specifications
  • Ext.Ø:
    127 mm (5")
  • Height:
    136.5 mm (5⅜")