ContactUsLinkComponent
Knife Sharpener, Multi-Edge Diamond Hone, Mortech
Knife Sharpener, Multi-Edge Diamond Hone, Mortech
Catalog # 76549-376
undefined
Knife Sharpener, Multi-Edge Diamond Hone, Mortech
Catalog # 76549-376
Supplier Number:  AH120
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Knife sharpener, multi-edge diamond hone
  • Cat. no.:
    76549-376
Specifications
About this item
This product has two stages: forming and fining for a razor sharp, longer lasting double bevel edge. It uses a 100% ultra-fine diamond abrasive. Patented, precision guides for sharper edges. Can be used to sharpen straight edges and serrated knives. Precise bevel angle control.
  • Suitable for left or right handed individuals
  • No assembly required