Scissors Sharpener, Electric Diamond Hone, Mortech
Product Details & Documents
A revolutionary development in scissor sharpening. Produces quality sharpness with unparalleled simplicity. For incredibly sharp, straight scissors. Sharpens from tip to handle without the need to disassemble scissor. 100% diamond abrasive in stages 1 and 2 for superior results.
- Two (2) stages, sharpening and honing, for a razor sharp, longer lasting double bevel edge
- Precise bevel angle control for a 20° edge
- Ready to use, left or right handed
- Safe for all quality scissors
- 120 V
Double bevel edge stays sharp longer. Never-tempered patented technology eliminates guesswork for sharper edges. Compact, easy, safe, and rugged design.
Ordering information: 1-year manufacturers warranty.
Specifications
- Description:Scissors sharpener, electric diamond hone
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)