Orders LinkContactUsLinkComponent
Forceps, Extracting, #69, Mortech
Forceps, Extracting, #69, Mortech
7.25"
Catalog # 76548-866
Supplier:   Mopec
Supplier #:  AB300
Forceps, Extracting, #69, Mortech
Catalog # 76548-866
Supplier:  Mopec
Forceps, Extracting, #69, Mortech
Forceps, Extracting, #69, Mortech
7.25"
Price
$107.12
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Post-mortem extracting forceps used for upper and lower tooth fragments and roots.
  • German grade stainless steel
  • American pattern handle
Specifications
  • Length:
    7.25"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)