Orders LinkContactUsLinkComponent
Forceps, Rubber Tipped, Mortech
Forceps, Rubber Tipped, Mortech
Supplier:   Mopec
Forceps, Rubber Tipped, Mortech
Supplier:  Mopec
Certificates
Forceps, Rubber Tipped, Mortech
Forceps, Rubber Tipped, Mortech
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Soft rubber tipped forceps allow for secure post-mortem dissection with minimal tissue damage or compression. An excellent option for techniques that require no metal exposure. Blue tips provide excellent contrast for specimen photography.
  • Standard grade stainless steel
  • Rubber rounded tips
Available in a variety of lengths.
NOTE: Do not autoclave. Rinse under water or wipe clean with any of our disinfecting cleaners.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • 6"
    Log in for availability
    Log in for availability
    Each
    Each - $39.77
    $39.77
    Each
       $39.77
    Supplier Number: AB060
  • 8"
    Log in for availability
    Log in for availability
    Each
    Each - $54.75
    $54.75
    Each
       $54.75
    Supplier Number: AB061
  • 10"
    Log in for availability
    Log in for availability
    Each
    Each - $67.22
    $67.22
    Each
       $67.22
    Supplier Number: AB062
  • 12"
    Log in for availability
    Log in for availability
    Each
    Each - $67.22
    $67.22
    Each
       $67.22
    Supplier Number: AB063
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.