Orders LinkContactUsLinkComponent
Forceps, Bayonet, Mortech
Forceps, Bayonet, Mortech
6.5"
Catalog # 76548-722
Supplier:   Mopec
Supplier #:  AB050
Forceps, Bayonet, Mortech
Catalog # 76548-722
Supplier:  Mopec
Forceps, Bayonet, Mortech
Forceps, Bayonet, Mortech
6.5"
Price
$204.41
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ergonomically designed for a more comfortable wrist position during post-mortem dissections. Offers speculum function for hollow cavity specimens such as uterus, bladder and colon.
  • Standard grade stainless steel
  • Off-set serrated tip
Specifications
  • Length:
    6.5"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.