Orders LinkContactUsLinkComponent
Forceps, Needle Core Biopsy, Mortech
Forceps, Needle Core Biopsy, Mortech
4.25"
Catalog # 76548-720
Supplier:   Mopec
Supplier #:  AB049
Forceps, Needle Core Biopsy, Mortech
Catalog # 76548-720
Supplier:  Mopec
Forceps, Needle Core Biopsy, Mortech
Forceps, Needle Core Biopsy, Mortech
4.25"
Price
$216.90
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Modified tip design to hold and hook all types of core biopsy tissue.
  • Standard grade stainless steel
Specifications
  • Length:
    4.25"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)