Orders LinkContactUsLinkComponent
Forceps, Iris Tissue, Straight, Serrated, Mortech
Forceps, Iris Tissue, Straight, Serrated, Mortech
4"
Catalog # 76548-692
Supplier:   Mopec
Supplier #:  AB035
Forceps, Iris Tissue, Straight, Serrated, Mortech
Catalog # 76548-692
Supplier:  Mopec
Forceps, Iris Tissue, Straight, Serrated, Mortech
Forceps, Iris Tissue, Straight, Serrated, Mortech
4"
Price
$42.29
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Serrated tip forceps suitable for delicate post-mortem dissection.
  • Standard grade stainless steel
  • Serrated pattern tips
  • Ribbed grip
Specifications
  • Length:
    4"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)