Orders LinkContactUsLinkComponent
Forceps, Adson Dressing, Mortech
Forceps, Adson Dressing, Mortech
4.75"
Catalog # 76548-676
Supplier:   Mopec
Supplier #:  AB027
Forceps, Adson Dressing, Mortech
Catalog # 76548-676
Supplier:  Mopec
Forceps, Adson Dressing, Mortech
Forceps, Adson Dressing, Mortech
4.75"
Price
$52.25
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Adson dressing forceps are designed to hold and manipulate delicate tissues and dressing materials such as cotton or gauze during any post-mortem dissection procedures.
  • Standard grade stainless steel
Specifications
  • Length:
    4.75"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)