Orders LinkContactUsLinkComponent
Scissors, Titanium, Mayo, Mortech
 :   Mopec
Scissors, Titanium, Mayo, Mortech
 :  Mopec
Scissors, Titanium, Mayo, Mortech
Scissors, Titanium, Mayo, Mortech
Titanium alloy scissors weigh approximately 25% less than stainless steel, making them the first choice for many technicians. Titanium alloy instruments keep edges sharp longer than traditional instruments. More stain resistant than stainless steel.
  • Standard grade titanium alloy
  • 25% lighter that stainless steel
Available with straight or curved edges. Do not use bleach or other caustic chemicals.
Product Family Options
  • Length
    Curvature
  • 6.75"
    Straight
    Each
    Each - $316.66
    $316.66
    Each
       $316.66
     : AA701
  • 6.75"
    Curved
    Each
    Each - $274.29
    $274.29
    Each
       $274.29
     : AA702
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)