Orders LinkContactUsLinkComponent
Dissecting Kit For Small Animals, Mortech
Dissecting Kit For Small Animals, Mortech
Catalog # 76548-514
Supplier:   Mopec
Supplier #:  AA320
Dissecting Kit For Small Animals, Mortech
Catalog # 76548-514
Supplier:  Mopec
Dissecting Kit For Small Animals, Mortech
Dissecting Kit For Small Animals, Mortech
Price
448,88 $
Log in for availability
Each
Product Details & Documents
This kit includes an array of instruments designed for every aspect of small, post-mortem dissection.
Delivery information: Kit contains one each of the following: dissection scissors, sharp/blunt, straight – AA008, Mayo dissection scissors, straight – AA020, iris scissors, straight – AA028, general dissection forceps, serrated tips – AB003, iris tissue forceps, fine point 4.5" – AB041, hemostatic Kelley forceps – AC003, hemostatic Rochester pean forceps, straight – AC006, double prong flesh hooks – AD009, section lifter single ended – AD042, blade handle #3 – AJ140, ruler.