Orders LinkContactUsLinkComponent
Scissors, Potts-Smith, Angled, Mortech
Scissors, Potts-Smith, Angled, Mortech
Supplier:   Mopec
Scissors, Potts-Smith, Angled, Mortech
Supplier:  Mopec
Certificates
Scissors, Potts-Smith, Angled, Mortech
Scissors, Potts-Smith, Angled, Mortech
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Angled tip for hard to reach, post-mortem dissection techniques.
  • Highest quality German stainless steel
  • Sharp tips
Available with 45 or 60° angled tips.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • 76548-480
    60° Angle
    60° Angle
    7.5"
    Log in for availability
    Log in for availability
    Each
    Each - $623.53
    $623.53
    Each
       $623.53
    Supplier Number: AA144
  • 76548-482
    45° Angle
    45° Angle
    7.5"
    Log in for availability
    Log in for availability
    Each
    Each - $623.53
    $623.53
    Each
       $623.53
    Supplier Number: AA145
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)