Orders LinkContactUsLinkComponent
Scissors, Mayo Dissection, Straight, Mortech
Scissors, Mayo Dissection, Straight, Mortech
Supplier:   Mopec
Scissors, Mayo Dissection, Straight, Mortech
Supplier:  Mopec
Certificates
Scissors, Mayo Dissection, Straight, Mortech
Scissors, Mayo Dissection, Straight, Mortech
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Slightly blunt tips assure controlled, precise and safe post-mortem dissection with minimal damage to surrounding tissue.
  • Stainless steel
  • Straight cutting edge
  • Blunt tips
Available in standard or premium grade.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • 76548-344
    Standard Grade
    Standard Grade
    5.5"
    Log in for availability
    Log in for availability
    Each
    Each - $64.72
    $64.72
    Each
       $64.72
    Supplier Number: AA019
  • 76548-346
    Standard Grade
    Standard Grade
    6.75"
    Log in for availability
    Log in for availability
    Each
    Each - $89.66
    $89.66
    Each
       $89.66
    Supplier Number: AA020
  • 76548-390
    Standard Grade
    Standard Grade
    9"
    Log in for availability
    Log in for availability
    Each
    Each - $107.12
    $107.12
    Each
       $107.12
    Supplier Number: AA049
  • 76548-438
    Premium Grade
    Premium Grade
    5.5"
    Log in for availability
    Log in for availability
    Each
    Each - $176.98
    $176.98
    Each
       $176.98
    Supplier Number: AA119
  • 76548-440
    Premium Grade
    Premium Grade
    6.75"
    Log in for availability
    Log in for availability
    Each
    Each - $194.46
    $194.46
    Each
       $194.46
    Supplier Number: AA120
  • 76548-504
    Premium Grade
    Premium Grade
    9"
    Log in for availability
    Log in for availability
    Each
    Each - $229.35
    $229.35
    Each
       $229.35
    Supplier Number: AA221
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)