ContactUsLinkComponent
Algae Survey Slide Set of 15
Algae Survey Slide Set of 15
  470339-616
undefined
Algae Survey Slide Set of 15
  470339-616
 :  UNSLALGE15
  • Description:
    Algae survey slide set of 15
  • Cat. no.:
    470339-616
Collection of popular Algae samples.
  • Glass prepared microscope slides
  • Plastic storage case
  • Key chart is included on lid
Glass prepared microscope slides, supplied in a plastic case. A collection of slides specific to the study of algae. Set of 15.
Set includes: Chlamydomonas, Protococcus, Anabaena, Oedogonium, Gloeocapsa, Vaucheria, Spirogyra Veg, Ectocarpus, Nostoc, Mixed desmids, Laminaria, Volvox, Mixed diatoms, Nemalion, Fucus.